siRNA Details
Gene Symbol: TUSC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172367.2 | NP_758955.2 | 1 | 443 | ||
GenBank mRNA | AB090231.1 | - | 1 | 443 | ||
BC142615.1 | - | 1 | 427 | 40147346 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TUSC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172367.2 | NP_758955.2 | 1 | 443 | ||
GenBank mRNA | AB090231.1 | - | 1 | 443 | ||
BC142615.1 | - | 1 | 427 | 40147346 |