siRNA Details
| Gene Symbol: PIP5KL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135219.1 | NP_001128691.1 | 8 | 782 | ||
| NM_173492.1 | NP_775763.1 | 3 | 338 | |||
| GenBank mRNA | BC042184.1 | - | 3 | 338 | 5163193 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

PIP5KL1| Gene Symbol: PIP5KL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135219.1 | NP_001128691.1 | 8 | 782 | ||
| NM_173492.1 | NP_775763.1 | 3 | 338 | |||
| GenBank mRNA | BC042184.1 | - | 3 | 338 | 5163193 | |