siRNA Details
| Gene Symbol: C1QC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114101.1 | NP_001107573.1 | 3 | 578 | ||
| NM_172369.3 | NP_758957.2 | 3 | 555 | |||
| GenBank mRNA | AK057792.1 | - | 3 | 570 | ||
| AK130613.1 | - | 3 | 553 | |||
| AK226152.1 | - | 1 | 3850 | |||
| BC009016.1 | - | 3 | 520 | 4212772 | ||
| DA849505.1 | - | 3 | 548 | |||

C1QC
Genomic Map