siRNA Details
| Gene Symbol: KRT78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173352.3 | NP_775487.2 | 1 | 411 | ||
| XM_011538010.1 | XP_011536312.1 | 1 | 411 | |||
| GenBank mRNA | AK096419.1 | - | 1 | 411 | ||
| AK290901.1 | - | 1 | 411 | |||
| AK298636.1 | - | 2 | 337 | |||
| DA667429.1 | - | 1 | 412 | |||

KRT78
Genomic Map