siRNA Details
| Gene Symbol: EIF4E3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134649.2 | NP_001128121.1 | 8 | 804 | ||
| NM_001134650.1 | NP_001128122.1 | 8 | 680 | |||
| NM_001134651.1 | NP_001128123.1 | 7 | 671 | |||
| NM_001282886.1 | NP_001269815.1 | 8 | 797 | |||
| NM_173359.4 | NP_775495.1 | 8 | 765 | |||
| XM_011533651.2 | XP_011531953.1 | 5 | 524 | |||
| XM_017006278.1 | XP_016861767.1 | 9 | 811 | |||
| XM_017006279.1 | XP_016861768.1 | 9 | 798 | |||
| XM_017006280.1 | XP_016861769.1 | 9 | 945 | |||
| XM_017006281.1 | XP_016861770.1 | 7 | 673 | |||
| XM_017006282.1 | XP_016861771.1 | 9 | 753 | |||
| GenBank mRNA | AK126999.1 | - | 7 | 553 | ||
| AK313656.1 | - | 8 | 765 | |||
| BC031289.2 | - | 6 | 488 | 5299499 | ||
| BC068443.1 | - | 8 | 797 | 4820931 | ||
| BX648727.1 | - | 4 | 285 | |||
| DR159502.1 | - | 7 | 670 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_025829, BC027014, AK005054, AK007268, AY628329.

EIF4E3
Genomic Map