siRNA Details
| Gene Symbol: MISP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173481.3 | NP_775752.1 | 2 | 481 | ||
| XM_011527685.2 | XP_011525987.1 | 2 | 526 | |||
| XM_011527686.2 | XP_011525988.1 | 2 | 619 | |||
| GenBank mRNA | BC042125.2 | - | 2 | 448 | 3848272 | |
| BC052236.1 | - | 2 | 468 | 5431360 | ||
| BX648389.1 | - | 2 | 462 | |||
| HY163020.1 | - | 2 | 483 | |||

MISP
Genomic Map