siRNA Details
| Gene Symbol: SPRR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173080.2 | NP_775103.1 | 2 | 439 | ||
| XM_017000482.1 | XP_016855971.1 | 3 | 1135 | |||
| GenBank mRNA | W77994.1 | - | 1 | 372 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPRR4| Gene Symbol: SPRR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173080.2 | NP_775103.1 | 2 | 439 | ||
| XM_017000482.1 | XP_016855971.1 | 3 | 1135 | |||
| GenBank mRNA | W77994.1 | - | 1 | 372 | ||