siRNA Details
| Gene Symbol: ARMC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282745.1 | NP_001269674.1 | 7, 8 | 824 | ||
| NM_001282746.1 | NP_001269675.1 | 7, 8 | 824 | |||
| NM_001282747.1 | NP_001269676.1 | 4, 5 | 335 | |||
| NM_173081.4 | NP_775104.2 | 7, 8 | 824 | |||
| XM_005252381.2 | XP_005252438.1 | 6, 7 | 645 | |||
| XM_011519350.2 | XP_011517652.1 | 7, 8 | 807 | |||
| XM_017015832.1 | XP_016871321.1 | 7, 8 | 807 | |||
| GenBank mRNA | AK057389.1 | - | 7, 8 | 767 | ||
| AK302104.1 | - | 4, 5 | 282 | |||
| AK302985.1 | - | 7, 8 | 811 | |||
| AY436643.1 | - | 7, 8 | 785 | |||
| BC039312.1 | - | 7, 8 | 817 | 5296302 | ||

ARMC3
Genomic Map