siRNA Details
| Gene Symbol: NKAPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027131.1 | 2 | 543 | |||
| GenBank mRNA | AK093895.1 | - | 2 | 509 | ||
| BC022359.1 | - | 2 | 483 | |||
| BC113730.1 | - | 2 | 482 | |||
| BC113732.1 | - | 2 | 482 | |||
| BX537594.1 | - | 1 | 4941 | |||

NKAPP1
Genomic Map