siRNA Details
| Gene Symbol: FAM71D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173526.3 | NP_775797.2 | 3, 4 | 457 | ||
| XM_011536500.1 | XP_011534802.1 | 3, 4 | 421 | |||
| XM_011536502.1 | XP_011534804.1 | 3, 4 | 356 | |||
| XM_011536503.2 | XP_011534805.1 | 2, 3 | 1398 | |||
| GenBank mRNA | AK093449.1 | - | 3, 4 | 421 | ||
| BC050401.1 | - | 3, 4 | 389 | 5745029 | ||
| DB056257.1 | - | 4, 5 | 516 | |||

FAM71D
Genomic Map