siRNA Details
| Gene Symbol: ZNF610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161425.1 | NP_001154897.1 | 5 | 763 | ||
| NM_001161426.1 | NP_001154898.1 | 5 | 650 | |||
| NM_173530.2 | NP_775801.2 | 5 | 619 | |||
| GenBank mRNA | AK093359.1 | - | 5 | 619 | ||
| AK290888.1 | - | 5 | 700 | |||
| BC105967.1 | - | 5 | 537 | 6154656 | ||

ZNF610
Genomic Map