siRNA Details
| Gene Symbol: ZNF100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173531.3 | NP_775802.2 | 3, 4 | 433 | ||
| GenBank mRNA | AK126551.1 | - | 3, 4 | 383 | ||
| AK308798.1 | - | 3, 4 | 354 | |||
| BC035579.1 | - | 3, 4 | 345 | 5576923 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF100| Gene Symbol: ZNF100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173531.3 | NP_775802.2 | 3, 4 | 433 | ||
| GenBank mRNA | AK126551.1 | - | 3, 4 | 383 | ||
| AK308798.1 | - | 3, 4 | 354 | |||
| BC035579.1 | - | 3, 4 | 345 | 5576923 | ||