siRNA Details
| Gene Symbol: FNDC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144937.1 | NP_001138409.1 | 6 | 1076 | ||
| GenBank mRNA | AK093157.1 | - | 2 | 402 | ||
| BC057762.1 | - | 2 | 449 | |||
| BC130421.1 | - | 2 | 402 | 40146452 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FNDC7| Gene Symbol: FNDC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144937.1 | NP_001138409.1 | 6 | 1076 | ||
| GenBank mRNA | AK093157.1 | - | 2 | 402 | ||
| BC057762.1 | - | 2 | 449 | |||
| BC130421.1 | - | 2 | 402 | 40146452 | ||