siRNA Details
| Gene Symbol: KLHDC8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173546.2 | NP_775817.1 | 6 | 1351 | ||
| XM_005264938.2 | XP_005264995.1 | 6 | 1837 | |||
| XM_005264940.4 | XP_005264997.1 | 6 | 1472 | |||
| XM_006713015.2 | XP_006713078.1 | 6 | 1867 | |||
| XM_006713016.2 | XP_006713079.1 | 6 | 1846 | |||
| GenBank mRNA | AK002164.1 | - | 2 | 1127 | ||
| AK294160.1 | - | 6 | 1200 | |||
| AY129011.1 | - | 6 | 1308 | |||
| BC013110.2 | - | 2 | 344 | |||
| BC039083.1 | - | 6 | 1301 | 5167173 | ||

KLHDC8B
Genomic Map