siRNA Details
| Gene Symbol: C3orf58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134470.1 | NP_001127942.1 | 2 | 492 | ||
| NM_173552.3 | NP_775823.1 | 2 | 1446 | |||
| XM_017005869.1 | XP_016861358.1 | 2 | 326 | |||
| GenBank mRNA | AK095161.1 | - | 2 | 508 | ||
| AK095400.1 | - | 2 | 352 | |||
| AK294020.1 | - | 2 | 390 | |||
| AK315082.1 | - | 2 | 1420 | |||
| AL832839.1 | - | 2 | 508 | |||
| BC037293.2 | - | 2 | 1173 | 5267770 | ||
| DC310421.1 | - | 2 | 390 | |||

C3orf58
Genomic Map