siRNA Details
| Gene Symbol: UNC5CL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173561.2 | NP_775832.2 | 2 | 461 | ||
| GenBank mRNA | AI791395.1 | - | 2 | 461 | ||
| AY510109.1 | - | 1 | 362 | |||
| BC035284.1 | - | 2 | 451 | 5189247 | ||
| JF681947.1 | - | 2 | 479 | |||

UNC5CL
Genomic Map