siRNA Details
| Gene Symbol: VKORC1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284342.1 | NP_001271271.1 | 2 | 737 | ||
| NM_173517.4 | NP_775788.2 | 3 | 847 | |||
| XM_011515830.2 | XP_011514132.1 | 4 | 2643 | |||
| XM_011515831.2 | XP_011514133.1 | 4 | 673 | |||
| GenBank mRNA | AK124688.1 | - | 2 | 500 | ||
| AK289522.1 | - | 3 | 760 | |||
| AK304077.1 | - | 2 | 641 | |||
| AY423045.1 | - | 3 | 766 | |||
| BC027734.1 | - | 3 | 847 | 5093832 | ||
| BX647631.1 | - | 3 | 780 | |||
| CR627471.1 | - | 2 | 535 | |||

VKORC1L1
Genomic Map