siRNA Details
| Gene Symbol: NKAPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027131.1 | 3 | 682 | |||
| GenBank mRNA | AK093895.1 | - | 3 | 648 | ||
| BC022359.1 | - | 3 | 622 | |||
| BC113730.1 | - | 3 | 621 | |||
| BC113732.1 | - | 3 | 621 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NKAPP1| Gene Symbol: NKAPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027131.1 | 3 | 682 | |||
| GenBank mRNA | AK093895.1 | - | 3 | 648 | ||
| BC022359.1 | - | 3 | 622 | |||
| BC113730.1 | - | 3 | 621 | |||
| BC113732.1 | - | 3 | 621 | |||