siRNA Details
| Gene Symbol: C10orf91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173541.2 | NP_775812.1 | 3 | 626 | ||
| GenBank mRNA | BC030794.1 | - | 3 | 647 | 3898116 | |
| BE909428.1 | - | 3 | 624 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C10orf91| Gene Symbol: C10orf91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173541.2 | NP_775812.1 | 3 | 626 | ||
| GenBank mRNA | BC030794.1 | - | 3 | 647 | 3898116 | |
| BE909428.1 | - | 3 | 624 | |||