siRNA Details
| Gene Symbol: NYAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173564.3 | NP_775835.2 | 4, 5 | 2102 | ||
| GenBank mRNA | AK094857.1 | - | 1, 2 | 213 | ||
| AK124773.1 | - | 4, 5 | 2102 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NYAP1| Gene Symbol: NYAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173564.3 | NP_775835.2 | 4, 5 | 2102 | ||
| GenBank mRNA | AK094857.1 | - | 1, 2 | 213 | ||
| AK124773.1 | - | 4, 5 | 2102 | |||