siRNA Details
| Gene Symbol: SPATA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304804.1 | NP_001291733.1 | 3 | 690 | ||
| NM_001304805.1 | NP_001291734.1 | 4 | 839 | |||
| NM_001304806.1 | NP_001291735.1 | 3 | 753 | |||
| NM_173499.4 | NP_775770.1 | 3 | 908 | |||
| XM_011521265.2 | XP_011519567.1 | 3 | 955 | |||
| GenBank mRNA | AY489187.1 | - | 3 | 872 | ||
| BC033979.2 | - | 3 | 845 | 5295377 | ||
| BC066292.1 | - | 3 | 867 | 5271997 | ||
| BC112390.1 | - | 2 | 1236 | 5168326 | ||

SPATA8
Genomic Map