siRNA Details
| Gene Symbol: MAGEB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173523.2 | NP_775794.2 | 2 | 977 | ||
| GenBank mRNA | AK097561.1 | - | 2 | 977 | ||
| BC067286.1 | - | 2 | 977 | 30386334 | ||
| BC074920.2 | - | 1 | 865 | 30915342 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAGEB6| Gene Symbol: MAGEB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173523.2 | NP_775794.2 | 2 | 977 | ||
| GenBank mRNA | AK097561.1 | - | 2 | 977 | ||
| BC067286.1 | - | 2 | 977 | 30386334 | ||
| BC074920.2 | - | 1 | 865 | 30915342 | ||