siRNA Details
| Gene Symbol: ZNF100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173531.3 | NP_775802.2 | 5 | 4043 | ||
| GenBank mRNA | BC035579.1 | - | 5 | 3948 | 5576923 | |
| BX640982.1 | - | 1 | 2751 | |||
| CN302052.1 | - | 1 | 188 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF100| Gene Symbol: ZNF100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173531.3 | NP_775802.2 | 5 | 4043 | ||
| GenBank mRNA | BC035579.1 | - | 5 | 3948 | 5576923 | |
| BX640982.1 | - | 1 | 2751 | |||
| CN302052.1 | - | 1 | 188 | |||