siRNA Details
| Gene Symbol: KLHDC8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173546.2 | NP_775817.1 | 6 | 1790 | ||
| XM_005264938.2 | XP_005264995.1 | 6 | 2276 | |||
| XM_005264940.4 | XP_005264997.1 | 6 | 1911 | |||
| XM_006713015.2 | XP_006713078.1 | 6 | 2306 | |||
| XM_006713016.2 | XP_006713079.1 | 6 | 2285 | |||
| GenBank mRNA | AK002164.1 | - | 2 | 1566 | ||
| AY129011.1 | - | 6 | 1747 | |||
| BC013110.2 | - | 2 | 783 | |||
| BC039083.1 | - | 6 | 1740 | 5167173 | ||

KLHDC8B
Genomic Map