siRNA Details
| Gene Symbol: C3orf58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134470.1 | NP_001127942.1 | 3 | 2111 | ||
| NM_173552.3 | NP_775823.1 | 3 | 3065 | |||
| XM_011512543.2 | XP_011510845.1 | 2 | 2650 | |||
| XM_017005869.1 | XP_016861358.1 | 3 | 1945 | |||
| GenBank mRNA | AK025119.1 | - | 1 | 673 | ||
| AK095063.1 | - | 1 | 1430 | |||
| AK095161.1 | - | 3 | 2127 | |||
| AK095400.1 | - | 3 | 1972 | |||
| AL832839.1 | - | 3 | 2126 | |||
| BC037293.2 | - | 3 | 2792 | 5267770 | ||

C3orf58
Genomic Map