siRNA Details
| Gene Symbol: FAM177A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079519.1 | NP_001072987.1 | 3, 4 | 558 | ||
| NM_001289022.1 | NP_001275951.1 | 2, 3 | 281 | |||
| NM_173607.3 | NP_775878.2 | 1, 2 | 228 | |||
| GenBank mRNA | AK096173.1 | - | 2, 3 | 169 | ||
| AL833508.1 | - | 1, 2 | 142 | |||
| BC029559.1 | - | 2, 3 | 150 | 5310901 | ||
| BG700755.1 | - | 2, 3 | 281 | |||
| BX640884.1 | - | 1, 2 | 231 | |||
| BX640984.1 | - | 1, 2 | 179 | |||
| CR749796.1 | - | 1, 2 | 228 | |||

FAM177A1
Genomic Map