siRNA Details
| Gene Symbol: ARL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317948.1 | NP_001304877.1 | 2 | 464 | ||
| NM_173664.5 | NP_775935.1 | 2 | 464 | |||
| XM_011534529.2 | XP_011532831.1 | 2 | 458 | |||
| XM_011534530.2 | XP_011532832.1 | 2 | 458 | |||
| XM_011534531.2 | XP_011532833.1 | 2 | 458 | |||
| XM_017009372.1 | XP_016864861.1 | 2 | 458 | |||
| GenBank mRNA | BC059361.1 | - | 2 | 464 | 30344341 | |
| DR001833.1 | - | 2 | 438 | |||

ARL10
Genomic Map