siRNA Details
| Gene Symbol: LOC100996634 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277339.1 | NP_001264268.1 | 1 | 155 | ||
| GenBank mRNA | AK094715.1 | - | 1 | 155 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100996634| Gene Symbol: LOC100996634 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277339.1 | NP_001264268.1 | 1 | 155 | ||
| GenBank mRNA | AK094715.1 | - | 1 | 155 | ||