siRNA Details
| Gene Symbol: GOLGA8T | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033933.1 | 8, 9 | 708 | |||
| Gene Symbol: GOLGA8H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282490.1 | NP_001269419.1 | 8, 9 | 686 | ||
| Gene Symbol: GOLGA8IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024074.1 | 7, 8 | 889 | |||
| GenBank mRNA | AK093104.1 | - | 7, 8 | 889 | ||
| Gene Symbol: GOLGA8J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282472.1 | NP_001269401.1 | 8, 9 | 688 | ||
| XM_006720651.3 | XP_006720714.1 | 8, 9 | 708 | |||
| XM_011521948.2 | XP_011520250.1 | 8, 9 | 1016 | |||
| XM_011521950.2 | XP_011520252.1 | 8, 9 | 1014 | |||
| Gene Symbol: GOLGA8K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282493.1 | NP_001269422.1 | 8, 9 | 688 | ||
| Gene Symbol: GOLGA8M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282468.1 | NP_001269397.1 | 8, 9 | 688 | ||
| XM_005268284.4 | XP_005268341.1 | 8, 9 | 1487 | |||
| XM_005268285.4 | XP_005268342.1 | 8, 9 | 1487 | |||
| XM_006720654.3 | XP_006720717.1 | 8, 9 | 631 | |||
| XM_011521951.2 | XP_011520253.1 | 9, 10 | 1056 | |||
| XM_011521952.2 | XP_011520254.1 | 8, 9 | 1487 | |||
| XM_011521953.2 | XP_011520255.1 | 8, 9 | 1487 | |||
| XM_011521954.2 | XP_011520256.1 | 8, 9 | 621 | |||
| Gene Symbol: GOLGA8N | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282494.1 | NP_001269423.1 | 8, 9 | 688 | ||
| Gene Symbol: GOLGA8O | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277308.1 | NP_001264237.1 | 8, 9 | 688 | ||
| XM_011521988.2 | XP_011520290.1 | 8, 9 | 1373 | |||
| XM_011521989.2 | XP_011520291.1 | 8, 9 | 1377 | |||
| GenBank mRNA | BQ477899.1 | - | 3, 4 | 218 | ||
| Gene Symbol: GOLGA8R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282484.1 | NP_001269413.1 | 8, 9 | 688 | ||

GOLGA8T
Genomic Map