siRNA Details
| Gene Symbol: RSPO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038633.3 | NP_001033722.1 | 8 | 2227 | ||
| NM_001242908.1 | NP_001229837.1 | 7 | 2048 | |||
| NM_001242909.1 | NP_001229838.1 | 7 | 1727 | |||
| NM_001242910.1 | NP_001229839.1 | 6 | 1859 | |||
| XM_006710583.3 | XP_006710646.1 | 6 | 1963 | |||
| GenBank mRNA | AK098225.1 | - | 7 | 1471 | ||
| BC114966.1 | - | 6 | 1647 | 40084277 | ||
| DQ165084.1 | - | 7 | 1792 | |||
| DQ165085.1 | - | 6 | 1603 | |||
| DQ318235.1 | - | 8 | 1938 | |||

RSPO1
Genomic Map