siRNA Details
| Gene Symbol: RIMKLA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173642.3 | NP_775913.2 | 4 | 770 | ||
| XM_017001082.1 | XP_016856571.1 | 5 | 829 | |||
| GenBank mRNA | BC039737.1 | - | 4 | 764 | 6021672 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RIMKLA| Gene Symbol: RIMKLA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173642.3 | NP_775913.2 | 4 | 770 | ||
| XM_017001082.1 | XP_016856571.1 | 5 | 829 | |||
| GenBank mRNA | BC039737.1 | - | 4 | 764 | 6021672 | |