siRNA Details
| Gene Symbol: DTWD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308081.1 | NP_001295010.1 | 3 | 241 | ||
| NM_173666.3 | NP_775937.1 | 3 | 462 | |||
| XM_011543338.2 | XP_011541640.2 | 3 | 1795 | |||
| XM_011543340.2 | XP_011541642.1 | 3 | 294 | |||
| GenBank mRNA | AK091296.1 | - | 3 | 402 | ||
| BC114508.1 | - | 3 | 398 | 40024023 | ||
| BC114955.1 | - | 3 | 398 | 40024022 | ||
| BX648369.1 | - | 3 | 241 | |||

DTWD2
Genomic Map