siRNA Details
| Gene Symbol: TMEM145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173633.2 | NP_775904.2 | 12 | 978 | ||
| XM_005258781.4 | XP_005258838.1 | 12 | 1020 | |||
| XM_011526791.2 | XP_011525093.1 | 12 | 972 | |||
| GenBank mRNA | AK075286.1 | - | 12 | 967 | ||

TMEM145
Genomic Map