siRNA Details
| Gene Symbol: FSIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173651.3 | NP_775922.3 | 11, 12 | 1248 | ||
| GenBank mRNA | AK092099.1 | - | 6, 7 | 579 | ||
| BC121088.1 | - | 7, 8 | 670 | |||
| BC121089.1 | - | 7, 8 | 670 | |||
| BC128052.1 | - | 7, 8 | 670 | |||
| BC148281.2 | - | 5, 6 | 456 | |||
| CR936597.1 | - | 6, 7 | 604 | |||

FSIP2
Genomic Map