siRNA Details
| Gene Symbol: CDC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134419.1 | NP_001127891.1 | 2 | 208 | ||
| NM_001134420.1 | NP_001127892.1 | 2 | 336 | |||
| NM_003503.3 | NP_003494.1 | 2 | 242 | |||
| XM_005271241.2 | XP_005271298.1 | 2 | 204 | |||
| XM_005271244.2 | XP_005271301.1 | 2 | 242 | |||
| XM_017002425.1 | XP_016857914.1 | 2 | 204 | |||
| XM_017002426.1 | XP_016857915.1 | 2 | 187 | |||
| GenBank mRNA | AB003698.1 | - | 2 | 209 | ||
| AB209337.1 | - | 1 | 918 | |||
| AF005209.1 | - | 1 | 77 | |||
| AF015592.1 | - | 2 | 222 | |||
| AK298963.1 | - | 2 | 238 | |||
| AK312726.1 | - | 2 | 298 | |||
| BC110526.1 | - | 2 | 165 | 40034178 | ||
| BC110527.1 | - | 2 | 199 | 40034179 | ||
| BC111044.1 | - | 2 | 222 | 5518616 | ||
| DA127303.1 | - | 2 | 336 | |||
| DA791490.1 | - | 2 | 242 | |||

CDC7
Genomic Map