siRNA Details
| Gene Symbol: C3orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308229.1 | NP_001295158.1 | 5 | 694 | ||
| NM_173657.2 | NP_775928.1 | 6 | 821 | |||
| XM_005247379.4 | XP_005247436.1 | 5 | 1666 | |||
| XM_011512710.2 | XP_011511012.1 | 4 | 619 | |||
| XM_011512711.2 | XP_011511013.1 | 5 | 798 | |||
| GenBank mRNA | AF115515.1 | - | 5 | 657 | ||
| AK055701.1 | - | 6 | 800 | |||
| AK289890.1 | - | 5 | 694 | |||
| AL832165.1 | - | 5 | 679 | |||
| BC042198.1 | - | 5 | 656 | |||
| BC064898.1 | - | 5 | 666 | 6008321 | ||

C3orf33
Genomic Map