siRNA Details
| Gene Symbol: RNF175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173662.2 | NP_775933.1 | 9 | 1339 | ||
| GenBank mRNA | BC034385.1 | - | 9 | 1340 | 5180678 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF175| Gene Symbol: RNF175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173662.2 | NP_775933.1 | 9 | 1339 | ||
| GenBank mRNA | BC034385.1 | - | 9 | 1340 | 5180678 | |