siRNA Details
Gene Symbol: LOC100996634 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277339.1 | NP_001264268.1 | 9 | 1444 | ||
XM_017010153.1 | XP_016865642.1 | 11 | 5741 | |||
XM_017010154.1 | XP_016865643.1 | 11 | 5741 | |||
GenBank mRNA | AK094715.1 | - | 9 | 1444 |
