siRNA Details
| Gene Symbol: SLC41A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173854.5 | NP_776253.3 | 2 | 1319 | ||
| XM_005245069.2 | XP_005245126.1 | 2 | 995 | |||
| GenBank mRNA | AJ514402.1 | - | 1 | 406 | ||
| AK128069.1 | - | 1 | 572 | |||
| AK314732.1 | - | 1 | 477 | |||
| AL831974.1 | - | 2 | 1155 | |||
| AL832001.1 | - | 2 | 1155 | |||
| AL832362.1 | - | 2 | 1155 | |||
| BC136303.1 | - | 1 | 800 | 9020290 | ||
| BX641054.1 | - | 2 | 1157 | |||
| BX648979.1 | - | 2 | 1193 | |||

SLC41A1
Genomic Map