siRNA Details
| Gene Symbol: TMEM167A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174909.4 | NP_777569.1 | 4 | 639 | ||
| GenBank mRNA | BC026285.1 | - | 4 | 529 | 4762531 | |
| BC028585.1 | - | 4 | 563 | 4836971 | ||
| BC070122.1 | - | 4 | 557 | |||
| BC107575.1 | - | 4 | 550 | 4282548 | ||
| BX647979.1 | - | 4 | 539 | |||

TMEM167A
Genomic Map