siRNA Details
| Gene Symbol: UBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174916.2 | NP_777576.1 | 2 | 260 | ||
| GenBank mRNA | AF525401.1 | - | 2 | 143 | ||
| AK056441.1 | - | 2 | 210 | |||
| AK292516.1 | - | 2 | 207 | |||
| AK302489.1 | - | 2 | 287 | |||
| AY061886.1 | - | 2 | 194 | |||
| BC113505.1 | - | 2 | 272 | 8322557 | ||
| BC113507.1 | - | 2 | 272 | 8322559 | ||

UBR1
Genomic Map