siRNA Details
| Gene Symbol: CCDC107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195200.1 | NP_001182129.1 | 5 | 599 | ||
| NM_001195201.1 | NP_001182130.1 | 5 | 579 | |||
| NM_001195217.1 | NP_001182146.1 | 5 | 571 | |||
| NM_174923.2 | NP_777583.2 | 5 | 599 | |||
| XM_005251403.4 | XP_005251460.1 | 5 | 582 | |||
| GenBank mRNA | AK075523.1 | - | 5 | 557 | ||
| BC018758.1 | - | 5 | 575 | 4907976 | ||
| BC051285.1 | - | 5 | 566 | 6586677 | ||
| BE903559.1 | - | 5 | 588 | |||
| DQ884403.1 | - | 5 | 509 | |||

CCDC107
Genomic Map