siRNA Details
| Gene Symbol: LINC00116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027063.1 | 7 | 1073 | |||
| GenBank mRNA | AF504645.1 | - | 7 | 1074 | ||
| BC064430.1 | - | 8 | 1505 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00116| Gene Symbol: LINC00116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027063.1 | 7 | 1073 | |||
| GenBank mRNA | AF504645.1 | - | 7 | 1074 | ||
| BC064430.1 | - | 8 | 1505 | |||