siRNA Details
| Gene Symbol: TMEM136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198670.1 | NP_001185599.1 | 2 | 567 | ||
| NM_001198671.1 | NP_001185600.1 | 1, 2 | 248 | |||
| NM_001198674.1 | NP_001185603.1 | 1, 2 | 248 | |||
| NM_174926.2 | NP_777586.1 | 2 | 567 | |||
| XM_017017327.1 | XP_016872816.1 | 1 | 3151 | |||
| GenBank mRNA | AK128040.1 | - | 1, 2 | 252 | ||
| AK294713.1 | - | 1, 2 | 76 | |||
| AK304086.1 | - | 2 | 400 | |||
| BC015232.1 | - | 2 | 389 | 3900115 | ||
| DA097438.1 | - | 1, 2 | 174 | |||

TMEM136
Genomic Map