siRNA Details
| Gene Symbol: DDX51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC040185.2 | - | 2, 3 | 539 | 4801253 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

DDX51| Gene Symbol: DDX51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC040185.2 | - | 2, 3 | 539 | 4801253 | |