siRNA Details
| Gene Symbol: LINC01554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026936.1 | 3 | 630 | |||
| GenBank mRNA | AK096140.1 | - | 4 | 529 | ||
| AK314737.1 | - | 3 | 630 | |||
| AY168789.1 | - | 4 | 509 | |||
| BC093643.1 | - | 2 | 139 | |||
| BC101577.1 | - | 2 | 139 | |||

LINC01554
Genomic Map