siRNA Details
| Gene Symbol: FAM126B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321618.1 | NP_001308547.1 | 6 | 606 | ||
| NM_001321619.1 | NP_001308548.1 | 7 | 817 | |||
| NM_001321621.1 | NP_001308550.1 | 6 | 719 | |||
| NM_001321622.1 | NP_001308551.1 | 8 | 935 | |||
| NM_001321623.1 | NP_001308552.1 | 7 | 704 | |||
| NM_001321624.1 | NP_001308553.1 | 6 | 606 | |||
| NM_001321625.1 | NP_001308554.1 | 7 | 817 | |||
| NM_001321626.1 | NP_001308555.1 | 7 | 837 | |||
| NM_001321627.1 | NP_001308556.1 | 8 | 760 | |||
| NM_001321628.1 | NP_001308557.1 | 8 | 873 | |||
| NM_001321629.1 | NP_001308558.1 | 8 | 760 | |||
| NM_173822.4 | NP_776183.1 | 7 | 704 | |||
| XM_006712443.3 | XP_006712506.1 | 6 | 719 | |||
| XM_011511002.2 | XP_011509304.1 | 7 | 734 | |||
| XM_017003879.1 | XP_016859368.1 | 8 | 915 | |||
| XM_017003880.1 | XP_016859369.1 | 7 | 1329 | |||
| XM_017003881.1 | XP_016859370.1 | 7 | 765 | |||
| XM_017003882.1 | XP_016859371.1 | 8 | 816 | |||
| XM_017003883.1 | XP_016859372.1 | 8 | 1159 | |||
| GenBank mRNA | AK124184.1 | - | 7 | 702 | ||
| AK124937.1 | - | 1 | 464 | |||
| AK315104.1 | - | 7 | 698 | |||
| AL832234.1 | - | 6 | 563 | |||
| AL832346.1 | - | 8 | 755 | |||
| AL833403.1 | - | 7 | 688 | |||
| BC039295.1 | - | 7 | 702 | 5267063 | ||

FAM126B
Genomic Map