siRNA Details
| Gene Symbol: C3orf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173824.3 | NP_776185.2 | 3 | 1046 | ||
| XM_005264745.4 | XP_005264802.1 | 3 | 1072 | |||
| GenBank mRNA | AK297719.1 | - | 3 | 696 | ||
| AK313547.1 | - | 3 | 787 | |||
| AL832398.1 | - | 3 | 1046 | |||
| BC024188.2 | - | 3 | 830 | 4824439 | ||
| KU178900.1 | - | 3 | 729 | |||
| KU178901.1 | - | 3 | 636 | |||

C3orf38
Genomic Map