siRNA Details
| Gene Symbol: GAS2L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303130.1 | NP_001290059.1 | 3 | 288 | ||
| NM_001303131.1 | NP_001290060.1 | 3 | 519 | |||
| NM_174942.2 | NP_777602.1 | 4 | 409 | |||
| XM_005268804.3 | XP_005268861.1 | 5 | 478 | |||
| XM_011538219.2 | XP_011536521.1 | 4 | 334 | |||
| XM_011538220.2 | XP_011536522.1 | 3 | 1669 | |||
| XM_011538221.2 | XP_011536523.1 | 3 | 519 | |||
| XM_017019206.1 | XP_016874695.1 | 5 | 552 | |||
| GenBank mRNA | AK095594.1 | - | 3 | 519 | ||
| AK315189.1 | - | 3 | 266 | |||
| BC043366.1 | - | 4 | 359 | 5104644 | ||
| BP872524.1 | - | 3 | 288 | |||

GAS2L3
Genomic Map