siRNA Details
| Gene Symbol: FPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005738.1 | NP_001005738.1 | 2 | 1843 | ||
| NM_001462.3 | NP_001453.1 | 2 | 1941 | |||
| XM_006723120.3 | XP_006723183.1 | 5 | 2674 | |||
| XM_017026542.1 | XP_016882031.1 | 4 | 2518 | |||
| GenBank mRNA | BC029125.2 | - | 1 | 1811 | 5180065 | |
| BC071722.1 | - | 2 | 2113 | 5212947 | ||
| D10922.1 | - | 2 | 1832 | |||
| M84562.1 | - | 2 | 2570 | |||
More Information
Set Membership: |
|

FPR2
Genomic Map